Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM234 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_133635.1 | 9 | 825 | |||
NR_133636.1 | 10 | 966 | ||||
XM_011541795.2 | XP_011540097.1 | 7 | 1838 | |||
XM_017001812.1 | XP_016857301.1 | 4 | 2445 | |||
XM_017001813.1 | XP_016857302.1 | 5 | 2193 | |||
XM_017001814.1 | XP_016857303.1 | 7 | 585 | |||
XM_017001815.1 | XP_016857304.1 | 7 | 917 | |||
XM_017001816.1 | XP_016857305.1 | 8 | 665 | |||
XM_017001817.1 | XP_016857306.1 | 6 | 1763 | |||
XM_017001818.1 | XP_016857307.1 | 7 | 1661 | |||
XM_017001819.1 | XP_016857308.1 | 9 | 1139 | |||
XM_017001820.1 | XP_016857309.1 | 7 | 1661 | |||
XM_017001821.1 | XP_016857310.1 | 9 | 807 | |||
XM_017001822.1 | XP_016857311.1 | 8 | 1263 | |||
XM_017001823.1 | XP_016857312.1 | 8 | 1205 | |||
XM_017001826.1 | XP_016857315.1 | 6 | 2010 | |||
XM_017001827.1 | XP_016857316.1 | 7 | 1908 | |||
XM_017001828.1 | XP_016857317.1 | 8 | 1178 | |||
XM_017001829.1 | XP_016857318.1 | 8 | 1510 | |||
XM_017001830.1 | XP_016857319.1 | 8 | 1178 | |||
GenBank mRNA | AF258548.1 | - | 8 | 1258 | ||
AK075260.1 | - | 3 | 2121 | |||
BC001256.1 | - | 10 | 928 | |||
BC038842.1 | - | 9 | 802 | 5742036 |