Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTFR1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099625.1 | NP_001093095.1 | 7 | 1320 | ||
NM_001099626.1 | NP_001093096.1 | 7 | 1372 | |||
NM_001099627.1 | NP_001093097.1 | 7 | 1211 | |||
NM_019557.5 | NP_062457.3 | 7 | 1418 | |||
GenBank mRNA | AB075880.1 | - | 7 | 1248 | ||
AF173891.1 | - | 7 | 1185 | |||
AK056721.1 | - | 6 | 2574 | |||
AK127164.1 | - | 3 | 1854 | |||
AK290744.1 | - | 6 | 1007 | |||
AL512766.1 | - | 7 | 1420 | |||
AY007100.1 | - | 7 | 1298 | |||
BC017175.2 | - | 7 | 1372 | 3162883 | ||
BX647921.1 | - | 7 | 1168 |