Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284395.1 | NP_001271324.1 | 8 | 971 | ||
NM_020212.1 | NP_064597.1 | 8 | 971 | |||
XM_006720614.1 | XP_006720677.1 | 8 | 957 | |||
XM_006720615.1 | XP_006720678.1 | 8 | 957 | |||
XM_006720618.2 | XP_006720681.1 | 8 | 957 | |||
XM_011521794.2 | XP_011520096.1 | 8 | 1250 | |||
XM_011521795.1 | XP_011520097.1 | 8 | 981 | |||
XM_011521796.2 | XP_011520098.1 | 8 | 957 | |||
XM_011521798.2 | XP_011520100.1 | 7 | 1241 | |||
XM_011521800.1 | XP_011520102.1 | 6 | 411 | |||
XM_011521802.2 | XP_011520104.1 | 8 | 957 | |||
XM_017022415.1 | XP_016877904.1 | 7 | 874 | |||
XM_017022416.1 | XP_016877905.1 | 5 | 341 | |||
GenBank mRNA | AK097525.1 | - | 8 | 939 | ||
AK302032.1 | - | 2 | 376 | |||
BC064626.1 | - | 8 | 971 | 5742386 |