Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020312.3 | NP_064708.1 | 6 | 754 | ||
GenBank mRNA | AF161444.1 | - | 6 | 688 | ||
AK290627.1 | - | 6 | 684 | |||
AL136884.1 | - | 1 | 188 | |||
BC001478.2 | - | 4 | 329 | |||
BC029341.1 | - | 6 | 688 | |||
BC054340.1 | - | 6 | 688 | 6514554 | ||
BC064946.1 | - | 6 | 689 | 6023461 |