Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317062.1 | NP_001303991.1 | 10 | 1744 | ||
NM_001317063.1 | NP_001303992.1 | 10 | 1702 | |||
NM_001317064.1 | NP_001303993.1 | 8 | 1516 | |||
NM_024102.3 | NP_077007.1 | 10 | 1708 | |||
NR_133654.1 | 9 | 1566 | ||||
GenBank mRNA | AB073603.1 | - | 10 | 1656 | ||
AB074171.1 | - | 5 | 1020 | |||
AK022860.1 | - | 10 | 1638 | |||
BC001679.2 | - | 10 | 1629 | 2822173 | ||
BC006477.1 | - | 10 | 1629 | 2822173 | ||
BC009411.1 | - | 11 | 1571 | 3614223 | ||
BC011778.2 | - | 10 | 1648 | 3941878 | ||
BC016946.1 | - | 10 | 1608 | 3845908 |