Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAA40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300800.1 | NP_001287729.1 | 4 | 431 | ||
NM_024771.3 | NP_079047.2 | 4 | 360 | |||
XM_006718689.3 | XP_006718752.1 | 4 | 347 | |||
XM_011545254.2 | XP_011543556.1 | 5 | 434 | |||
XM_011545255.2 | XP_011543557.1 | 5 | 434 | |||
GenBank mRNA | AK023910.1 | - | 4 | 329 | ||
AK299041.1 | - | 4 | 425 | |||
AK300451.1 | - | 3 | 281 | |||
AK303557.1 | - | 3 | 281 | |||
BC041617.1 | - | 4 | 289 | 5921480 | ||
BC052298.1 | - | 4 | 289 | 6292760 | ||
BX647309.1 | - | 3 | 1386 | |||
DA649381.1 | - | 4 | 365 | |||
DC307984.1 | - | 4 | 514 | |||
HY009487.1 | - | 4 | 431 | |||
JF263566.1 | - | 4 | 329 |