Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NAA40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300800.1 | NP_001287729.1 | 8 | 827 | ||
NM_024771.3 | NP_079047.2 | 8 | 756 | |||
XM_006718689.3 | XP_006718752.1 | 8 | 653 | |||
XM_011545254.2 | XP_011543556.1 | 9 | 830 | |||
XM_011545255.2 | XP_011543557.1 | 9 | 740 | |||
GenBank mRNA | AK023910.1 | - | 8 | 725 | ||
AK299041.1 | - | 8 | 821 | |||
AK300451.1 | - | 7 | 677 | |||
AK303557.1 | - | 6 | 587 | |||
BC024160.2 | - | 1 | 13 | |||
BC041617.1 | - | 8 | 685 | 5921480 | ||
BC052298.1 | - | 8 | 685 | 6292760 | ||
BX647309.1 | - | 7 | 1782 | |||
JF263566.1 | - | 8 | 725 |