Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002292.3 | NP_001002292.3 | 8 | 1334 | ||
NM_001193334.1 | NP_001180263.1 | 7 | 1067 | |||
NM_024911.6 | NP_079187.3 | 8 | 1395 | |||
XM_011542191.2 | XP_011540493.1 | 8 | 1318 | |||
XM_011542192.2 | XP_011540494.1 | 8 | 1328 | |||
XM_017002390.1 | XP_016857879.1 | 8 | 1162 | |||
GenBank mRNA | AK026744.1 | - | 5 | 476 | ||
AK074984.1 | - | 8 | 1317 | |||
AK301613.1 | - | 7 | 927 | |||
AK309779.1 | - | 9 | 1274 | |||
AY359035.1 | - | 8 | 1377 | |||
BC007211.1 | - | 7 | 933 | |||
BC110826.1 | - | 8 | 1296 | 6183768 | ||
BC137109.1 | - | 8 | 1192 | 9021106 | ||
BC137113.1 | - | 8 | 1192 | 9021110 | ||
BX538320.1 | - | 8 | 1334 | |||
BX648748.1 | - | 7 | 1065 | |||
DQ323735.1 | - | 8 | 1364 |