Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAF3IP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320143.1 | NP_001307072.1 | 17 | 2221 | ||
NM_001320144.1 | NP_001307073.1 | 17 | 2082 | |||
NM_025228.3 | NP_079504.2 | 17 | 2142 | |||
XM_005273279.4 | XP_005273336.1 | 10 | 2855 | |||
XM_005273280.2 | XP_005273337.1 | 10 | 1118 | |||
XM_017002399.1 | XP_016857888.1 | 16 | 2066 | |||
GenBank mRNA | AK092905.1 | - | 3 | 2491 | ||
AK122792.1 | - | 17 | 2125 | |||
AK225762.1 | - | 17 | 2037 | |||
AL049667.1 | - | 17 | 1935 | |||
BX648450.1 | - | 17 | 2121 |