Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3BP5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322462.1 | NP_001309391.1 | 5 | 1324 | ||
NM_001322463.1 | NP_001309392.1 | 3 | 699 | |||
NM_001322464.1 | NP_001309393.1 | 4 | 1150 | |||
NM_030645.1 | NP_085148.1 | 4 | 820 | |||
XM_017002404.1 | XP_016857893.1 | 3 | 1023 | |||
XM_017002405.1 | XP_016857894.1 | 3 | 802 | |||
XM_017002406.1 | XP_016857895.1 | 5 | 776 | |||
XM_017002407.1 | XP_016857896.1 | 3 | 708 | |||
GenBank mRNA | AB051507.1 | - | 3 | 527 | ||
AK056382.1 | - | 4 | 820 | |||
AK125138.1 | - | 4 | 1497 | |||
AK125837.1 | - | 5 | 1593 | |||
AK298698.1 | - | 3 | 174 | |||
AL136569.1 | - | 3 | 407 | |||
AY358453.1 | - | 4 | 742 | |||
BC010871.1 | - | 1 | 29 | |||
BC017254.2 | - | 3 | 309 | 3051344 |