Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISG20L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303095.1 | NP_001290024.1 | 3 | 1138 | ||
XM_005245516.4 | XP_005245573.1 | 3 | 1106 | |||
XM_005245518.4 | XP_005245575.1 | 2 | 653 | |||
XM_006711560.3 | XP_006711623.1 | 3 | 1276 | |||
XM_006711561.3 | XP_006711624.1 | 3 | 1165 | |||
GenBank mRNA | AK021901.1 | - | 2 | 318 | ||
AK022733.1 | - | 2 | 1196 | |||
AK095697.1 | - | 2 | 1718 | |||
AK311586.1 | - | 2 | 698 | |||
AY391837.1 | - | 2 | 1124 | |||
BC000575.2 | - | 3 | 1118 | 3162193 |