Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC88B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032251.5 | NP_115627.6 | 26 | 4410 | ||
XM_006718519.3 | XP_006718582.1 | 26 | 4280 | |||
XM_011544957.2 | XP_011543259.1 | 25 | 4518 | |||
XM_011544959.2 | XP_011543261.1 | 25 | 4660 | |||
XM_011544960.2 | XP_011543262.1 | 25 | 4184 | |||
XM_011544962.2 | XP_011543264.1 | 12 | 2140 | |||
GenBank mRNA | AK074126.1 | - | 19 | 4064 | ||
AK090436.1 | - | 24 | 4734 | |||
AK126728.1 | - | 5 | 1188 | |||
AL136799.1 | - | 3 | 371 | |||
AY491401.1 | - | 9 | 1131 | |||
BC030194.1 | - | 4 | 437 | |||
BC040232.1 | - | 13 | 1825 | |||
BC140822.1 | - | 26 | 4399 | 9021690 | ||
BC140824.1 | - | 26 | 4399 | 9021692 |