Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SGIP1 SIRNA Assays ›
Gene Symbol:
SGIP1
Gene Name:
SH3 domain GRB2 like endophilin interacting protein 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 66533272 - 66751139 on Build GRCh38
UniGene ID:
Hs.132121
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
123661
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SGIP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308203.1
NP_001295132.1
11
808
NM_032291.3
NP_115667.2
13
1090
XM_005271264.3
XP_005271321.1
13
924
XM_005271268.3
XP_005271325.1
12
840
XM_005271270.4
XP_005271327.1
11
1147
XM_006710961.2
XP_006711024.1
13
924
XM_006710966.2
XP_006711029.1
13
924
XM_006710967.2
XP_006711030.1
13
912
XM_006710969.2
XP_006711032.1
12
840
XM_006710971.2
XP_006711034.1
13
924
XM_006710972.2
XP_006711035.1
13
924
XM_006710973.2
XP_006711036.1
11
900
XM_006710974.2
XP_006711037.1
10
816
XM_011542291.1
XP_011540593.1
13
1087
XM_011542292.1
XP_011540594.1
13
1087
XM_011542293.1
XP_011540595.1
10
816
XM_017002505.1
XP_016857994.1
13
912
XM_017002506.1
XP_016857995.1
14
1035
XM_017002507.1
XP_016857996.1
13
1087
XM_017002508.1
XP_016857997.1
14
1033
XM_017002509.1
XP_016857998.1
11
888
XM_017002510.1
XP_016857999.1
12
1011
XM_017002511.1
XP_016858000.1
14
1035
XM_017002512.1
XP_016858001.1
14
908
XM_017002513.1
XP_016858002.1
13
1087
XM_017002514.1
XP_016858003.1
12
840
XM_017002515.1
XP_016858004.1
10
816
XM_017002516.1
XP_016858005.1
11
939
XM_017002517.1
XP_016858006.1
14
1034
XM_017002518.1
XP_016858007.1
13
962
XM_017002519.1
XP_016858008.1
13
987
XM_017002520.1
XP_016858009.1
11
828
XM_017002521.1
XP_016858010.1
12
840
XM_017002522.1
XP_016858011.1
13
962
XM_017002523.1
XP_016858012.1
13
986
XM_017002524.1
XP_016858013.1
12
1004
XM_017002525.1
XP_016858014.1
12
1003
XM_017002526.1
XP_016858015.1
11
888
XM_017002527.1
XP_016858016.1
12
912
XM_017002528.1
XP_016858017.1
11
932
XM_017002529.1
XP_016858018.1
13
924
XM_017002530.1
XP_016858019.1
12
900
XM_017002531.1
XP_016858020.1
11
900
XM_017002532.1
XP_016858021.1
11
828
XM_017002533.1
XP_016858022.1
12
1002
XM_017002534.1
XP_016858023.1
10
816
XM_017002535.1
XP_016858024.1
11
930
XM_017002536.1
XP_016858025.1
12
840
XM_017002537.1
XP_016858026.1
11
929
GenBank mRNA
AB210039.1
-
13
872
AK090697.1
-
5
365
AK298970.1
-
11
777
AK299941.1
-
12
851
AL136561.1
-
13
764
BC040516.1
-
14
837
5275011
BX640813.1
-
11
785
CR749541.1
-
11
1002
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
endocytosis
positive regulation of receptor-mediated endocytosis
membrane tubulation
protein binding
phospholipid binding
microtubule binding
SH3 domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-tbml7:80/10.239.166.229:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0