Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014765.1 | NP_001014765.1 | 7 | 1974 | ||
NM_001304790.1 | NP_001291719.1 | 6 | 1507 | |||
NM_001304791.1 | NP_001291720.1 | 6 | 1713 | |||
NM_033182.5 | NP_149438.2 | 6 | 1602 | |||
NM_183412.2 | NP_904319.1 | 5 | 1588 | |||
NM_183413.2 | NP_904320.1 | 5 | 1477 | |||
NR_130909.1 | 5 | 1343 | ||||
XM_005263535.1 | XP_005263592.1 | 6 | 1544 | |||
XM_005263536.4 | XP_005263593.1 | 7 | 1938 | |||
XM_005263537.1 | XP_005263594.1 | 6 | 1684 | |||
XM_011542435.1 | XP_011540737.1 | 7 | 2006 | |||
XM_017002842.1 | XP_016858331.1 | 7 | 1455 | |||
XM_017002843.1 | XP_016858332.1 | 7 | 1486 | |||
GenBank mRNA | AK055344.1 | - | 5 | 1588 | ||
AY040878.1 | - | 6 | 1558 | |||
BC007832.2 | - | 5 | 1424 | 4054414 | ||
CR749368.1 | - | 7 | 1963 |