Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322923.1 | NP_001309852.1 | 6 | 2893 | ||
NM_001322924.1 | NP_001309853.1 | 5 | 2821 | |||
NM_052862.3 | NP_443094.3 | 7 | 2939 | |||
GenBank mRNA | AF545852.1 | - | 7 | 3083 | ||
AK025425.1 | - | 2 | 2277 | |||
AK124221.1 | - | 1 | 2661 | |||
AL833211.1 | - | 5 | 2618 | |||
BC013186.1 | - | 2 | 1988 | |||
BC072399.1 | - | 2 | 2231 | |||
BC098426.1 | - | 2 | 2104 |