Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080659.2 | NP_542390.2 | 4 | 285 | ||
GenBank mRNA | AK057948.1 | - | 4 | 223 | ||
BC008501.1 | - | 4 | 249 | 4294167 | ||
BQ102454.1 | - | 4 | 285 |
Gene Symbol: HSPB2-C11orf52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037651.1 | 5 | 672 | |||
GenBank mRNA | BC110872.1 | - | 5 | 667 | 6193385 |