Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002927.4 | NP_002918.1 | 7 | 1025 | ||
NM_144766.2 | NP_658912.1 | 6 | 985 | |||
GenBank mRNA | AF030107.1 | - | 7 | 1033 | ||
BC016667.1 | - | 6 | 951 | 4340668 | ||
BC056866.1 | - | 7 | 1024 | 4419604 |