Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA5B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039500.2 | NP_001034589.2 | 21 | 3321 | ||
XM_006710360.3 | XP_006710423.2 | 18 | 2596 | |||
XM_011540683.2 | XP_011538985.1 | 21 | 3283 | |||
XM_011540684.2 | XP_011538986.1 | 21 | 3408 | |||
XM_011540685.2 | XP_011538987.1 | 21 | 3420 | |||
XM_011540688.2 | XP_011538990.1 | 20 | 3208 | |||
XM_011540690.2 | XP_011538992.1 | 20 | 3106 | |||
XM_011540691.2 | XP_011538993.1 | 20 | 3097 | |||
XM_011540692.2 | XP_011538994.1 | 21 | 3277 | |||
XM_011540697.2 | XP_011538999.1 | 15 | 2654 | |||
XM_017000305.1 | XP_016855794.1 | 15 | 2400 | |||
XM_017000306.1 | XP_016855795.1 | 16 | 2561 | |||
GenBank mRNA | AK057346.1 | - | 11 | 1605 | ||
AK295797.1 | - | 21 | 3305 | |||
BC101378.1 | - | 10 | 1401 | |||
BC101379.2 | - | 9 | 1326 | |||
BC101380.1 | - | 10 | 1401 | |||
BC101381.1 | - | 10 | 1401 |