Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: WDR66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178003.1 | NP_001171474.1 | 4 | 1046 | ||
NM_144668.5 | NP_653269.3 | 4 | 1033 | |||
GenBank mRNA | AA853421.1 | - | 1 | 58 | ||
AK097102.1 | - | 3 | 787 | |||
AL833930.1 | - | 2 | 321 | |||
BC028421.2 | - | 4 | 1031 | 4826893 | ||
BC036233.1 | - | 4 | 1026 | 5295406 |