Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144698.4 | NP_653299.4 | 4 | 413 | ||
GenBank mRNA | AK091120.1 | - | 4 | 378 | ||
AK294308.1 | - | 2 | 122 | |||
AK296861.1 | - | 4 | 378 | |||
BC101618.1 | - | 4 | 360 | 8069124 | ||
BC101620.1 | - | 4 | 360 | 8069126 | ||
DA399526.1 | - | 4 | 413 |