Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001280799.1 | NP_001267728.1 | 11 | 2876 | ||
NM_144698.4 | NP_653299.4 | 13 | 3146 | |||
GenBank mRNA | AK057853.1 | - | 4 | 1036 | ||
AK091120.1 | - | 13 | 3111 | |||
AK294308.1 | - | 11 | 2855 |