Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCSAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145257.4 | NP_660300.3 | 4 | 735 | ||
GenBank mRNA | AK123465.1 | - | 3 | 524 | ||
AK291437.1 | - | 3 | 696 | |||
BC015419.1 | - | 3 | 709 | 4396825 | ||
BC060777.1 | - | 4 | 1640 | |||
BC071609.1 | - | 4 | 781 | 5264741 |