Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCSAML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281834.1 | NP_001268763.1 | 7 | 1362 | ||
NM_001281835.1 | NP_001268764.1 | 6 | 1246 | |||
NM_001281836.1 | NP_001268765.1 | 6 | 1130 | |||
NM_001281837.1 | NP_001268766.1 | 6 | 1111 | |||
NM_001281838.1 | NP_001268767.1 | 5 | 1070 | |||
NM_001281853.1 | NP_001268782.1 | 7 | 1215 | |||
NM_145278.4 | NP_660321.1 | 5 | 1015 | |||
XM_017000399.1 | XP_016855888.1 | 6 | 1130 | |||
GenBank mRNA | AK123798.1 | - | 5 | 926 | ||
AK124520.1 | - | 7 | 1305 | |||
AK126682.1 | - | 6 | 1184 | |||
AK307081.1 | - | 5 | 1051 | |||
AK307082.1 | - | 6 | 1096 | |||
BC024174.1 | - | 5 | 915 | 4109064 |