Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RAB42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193532.1 | NP_001180461.1 | 2 | 498 | ||
NM_152304.1 | NP_689517.1 | 2 | 448 | |||
XM_017000226.1 | XP_016855715.1 | 2 | 519 | |||
GenBank mRNA | AK312176.1 | - | 2 | 385 | ||
BC033175.1 | - | 2 | 448 | 4561064 | ||
CX788289.1 | - | 2 | 505 |