Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152372.3 | NP_689585.3 | 26 | 3438 | ||
GenBank mRNA | AK128233.1 | - | 18 | 2498 | ||
BC067101.1 | - | 26 | 3448 | 5928128 | ||
BX537911.1 | - | 25 | 3162 | |||
BX647453.1 | - | 26 | 3399 | |||
BX647455.1 | - | 26 | 3438 | |||
BX647644.1 | - | 26 | 3399 |