Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LMNTD1 SIRNA Assays ›
Gene Symbol:
LMNTD1
Gene Name:
lamin tail domain containing 1
Gene Aliases:
IFLTD1, PAS1C1
Chromosome Location:
Chr.12: 25476082 - 25648604 on Build GRCh38
UniGene ID:
Hs.44647
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
124074
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LMNTD1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145727.2
NP_001139199.1
5
598
NM_001145728.2
NP_001139200.1
7
1084
NM_001145729.1
NP_001139201.1
6
964
NM_001256266.1
NP_001243195.1
7
996
NM_152590.3
NP_689803.2
6
965
XM_011520575.1
XP_011518877.1
9
1190
XM_011520576.2
XP_011518878.1
9
1190
XM_011520577.2
XP_011518879.1
9
1600
XM_011520578.2
XP_011518880.1
9
1213
XM_011520579.2
XP_011518881.1
9
1190
XM_011520580.1
XP_011518882.1
8
1070
XM_011520582.1
XP_011518884.1
9
1202
XM_011520583.1
XP_011518885.1
8
1071
XM_011520584.1
XP_011518886.1
6
878
XM_011520585.2
XP_011518887.1
6
801
XM_011520587.1
XP_011518889.1
6
574
XM_011520588.1
XP_011518890.1
5
496
XM_017018891.1
XP_016874380.1
8
1022
XM_017018892.1
XP_016874381.1
8
1092
XM_017018893.1
XP_016874382.1
7
901
XM_017018894.1
XP_016874383.1
10
1284
GenBank mRNA
AK093323.1
-
6
926
AK296816.1
-
5
590
AK302292.1
-
7
1084
AK310411.1
-
6
962
BC037957.1
-
7
965
5744078
BP359528.1
-
4
417
DW009862.1
-
2
99
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cell proliferation
structural molecule activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-cnw2s:80/10.239.163.240:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0