Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152608.3 | NP_689821.3 | 3 | 381 | ||
GenBank mRNA | AK127780.1 | - | 2 | 311 | ||
AK291008.1 | - | 3 | 344 | |||
AK308441.1 | - | 3 | 358 | |||
BC071563.1 | - | 3 | 359 | 5261192 | ||
BX648319.1 | - | 3 | 339 | |||
CB140971.1 | - | 3 | 381 |