Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319196.1 | NP_001306125.1 | 3 | 402 | ||
NM_001319197.1 | NP_001306126.1 | 3 | 399 | |||
NM_001319198.1 | NP_001306127.1 | 3 | 303 | |||
NM_001319201.1 | NP_001306130.1 | 3 | 357 | |||
NM_002964.4 | NP_002955.2 | 3 | 385 | |||
GenBank mRNA | AA401538.1 | - | 3 | 300 | ||
AK291328.1 | - | 3 | 270 | |||
BC005928.2 | - | 2 | 237 | 4246359 | ||
BP209293.1 | - | 3 | 287 | |||
BT007378.1 | - | 2 | 215 | |||
BU935536.1 | - | 3 | 387 | |||
CR407674.1 | - | 2 | 215 | |||
X06234.1 | - | 3 | 271 | |||
Y00278.1 | - | 3 | 266 |