Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM76A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143912.1 | NP_001137384.1 | 10 | 1992 | ||
NM_001143913.1 | NP_001137385.1 | 9 | 1905 | |||
NM_001143914.1 | NP_001137386.1 | 8 | 1803 | |||
NM_001143915.1 | NP_001137387.1 | 7 | 1650 | |||
NM_152660.2 | NP_689873.1 | 9 | 1890 | |||
GenBank mRNA | AK098318.1 | - | 8 | 1752 | ||
AK301672.1 | - | 7 | 1635 | |||
BC025768.1 | - | 9 | 1890 | 5211010 |