Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TCTEX1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152665.2 | NP_689878.2 | 4 | 408 | ||
XM_011540933.2 | XP_011539235.1 | 5 | 795 | |||
XM_017000566.1 | XP_016856055.1 | 4 | 704 | |||
GenBank mRNA | AK098192.1 | - | 4 | 406 | ||
AK307100.1 | - | 6 | 500 | |||
BC106885.2 | - | 4 | 373 | 40029630 | ||
DA520308.1 | - | 4 | 408 |