Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152760.2 | NP_689973.2 | 6 | 673 | ||
XM_006718488.3 | XP_006718551.1 | 6 | 976 | |||
XM_017017470.1 | XP_016872959.1 | 6 | 951 | |||
XM_017017471.1 | XP_016872960.1 | 7 | 1135 | |||
XM_017017472.1 | XP_016872961.1 | 5 | 1337 | |||
XM_017017473.1 | XP_016872962.1 | 5 | 1337 | |||
GenBank mRNA | AK055496.1 | - | 6 | 633 | ||
AK289587.1 | - | 6 | 639 | |||
AK299512.1 | - | 5 | 743 | |||
AK309789.1 | - | 4 | 889 | |||
AL833261.1 | - | 6 | 659 | |||
BC040981.2 | - | 6 | 649 | 4797976 | ||
BC045563.1 | - | 6 | 622 | 4815203 |