Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKNAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152763.4 | NP_689976.2 | 2 | 797 | ||
NR_049760.1 | 3 | 938 | ||||
GenBank mRNA | AK095517.1 | - | 3 | 924 | ||
AK313772.1 | - | 2 | 777 | |||
AL832216.1 | - | 2 | 787 | |||
BC030647.2 | - | 2 | 777 | |||
BC150642.1 | - | 1 | 552 | 9057012 |