Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDELC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153705.4 | NP_714916.3 | 2 | 425 | ||
XM_011542621.2 | XP_011540923.1 | 2 | 398 | |||
XM_011542623.2 | XP_011540925.1 | 2 | 392 | |||
GenBank mRNA | AF533708.1 | - | 2 | 384 | ||
AK125519.1 | - | 1 | 207 | |||
AK313534.1 | - | 2 | 287 | |||
AY358616.1 | - | 1 | 494 | |||
BC036526.2 | - | 2 | 379 | 5266665 | ||
DN999478.1 | - | 2 | 433 |