Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HARBI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173811.3 | NP_776172.1 | 3 | 1282 | ||
XM_011520025.1 | XP_011518327.1 | 4 | 1366 | |||
XM_011520026.2 | XP_011518328.1 | 4 | 1343 | |||
XM_011520027.1 | XP_011518329.1 | 4 | 1377 | |||
XM_011520028.2 | XP_011518330.1 | 4 | 1352 | |||
XM_011520029.2 | XP_011518331.1 | 4 | 1638 | |||
XM_017017602.1 | XP_016873091.1 | 4 | 1482 | |||
XM_017017603.1 | XP_016873092.1 | 4 | 1397 | |||
XM_017017604.1 | XP_016873093.1 | 3 | 1293 | |||
XM_017017605.1 | XP_016873094.1 | 3 | 1554 | |||
XM_017017606.1 | XP_016873095.1 | 3 | 1309 | |||
GenBank mRNA | AK057237.1 | - | 3 | 1282 | ||
BC036925.1 | - | 3 | 1214 | 5225925 | ||
BM768489.1 | - | 1 | 160 |