Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPAS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318804.1 | NP_001305733.1 | 6 | 1147 | ||
NM_178864.3 | NP_849195.2 | 7 | 1257 | |||
XM_017017537.1 | XP_016873026.1 | 9 | 2696 | |||
XM_017017538.1 | XP_016873027.1 | 3 | 409 | |||
GenBank mRNA | AB049469.1 | - | 7 | 1182 | ||
AK094025.1 | - | 4 | 562 | |||
AK096253.1 | - | 7 | 1257 | |||
BC105001.1 | - | 7 | 1170 | 8144004 | ||
BC105003.1 | - | 7 | 1170 | 8144006 | ||
BC143630.1 | - | 6 | 1060 |