Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181644.4 | NP_857595.3 | 10 | 1780 | ||
XM_011509229.2 | XP_011507531.1 | 9 | 1519 | |||
GenBank mRNA | AK057733.1 | - | 6 | 850 | ||
AK090831.1 | - | 4 | 507 | |||
AK091896.1 | - | 10 | 1721 | |||
AK127321.1 | - | 2 | 1416 | |||
AK293816.1 | - | 9 | 1709 | |||
AK304109.1 | - | 9 | 1585 | |||
AL713676.1 | - | 8 | 1086 | |||
AY358107.1 | - | 8 | 1167 | |||
BC036549.2 | - | 10 | 1730 | 5264261 |