Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC14L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039573.2 | NP_001034662.2 | 4 | 402 | ||
NM_001143998.1 | NP_001137470.1 | 4 | 402 | |||
NM_001143999.1 | NP_001137471.1 | 4 | 398 | |||
NM_001144001.1 | NP_001137473.1 | 2 | 190 | |||
NM_001204408.1 | NP_001191337.1 | 6 | 747 | |||
NM_001204410.1 | NP_001191339.1 | 5 | 440 | |||
NM_003003.3 | NP_002994.3 | 4 | 349 | |||
GenBank mRNA | AB208811.1 | - | 4 | 381 | ||
AK290913.1 | - | 4 | 295 | |||
AK293206.1 | - | 2 | 190 | |||
BC136523.1 | - | 4 | 237 | 9020513 | ||
BC136525.1 | - | 4 | 237 | 9020515 | ||
BC142979.1 | - | 4 | 293 | 8860414 | ||
BX537448.1 | - | 4 | 303 | |||
D67029.1 | - | 4 | 385 | |||
DA024354.1 | - | 4 | 340 | |||
DB040556.1 | - | 4 | 402 | |||
DB040580.1 | - | 2 | 190 | |||
FM995495.1 | - | 6 | 641 | |||
FM995496.1 | - | 5 | 438 | |||
FM995497.1 | - | 5 | 537 |