Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G4E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206670.1 | NP_001193599.1 | 20 | 4244 | ||
XM_011521237.2 | XP_011519539.1 | 20 | 4154 | |||
GenBank mRNA | AK127558.1 | - | 10 | 3533 | ||
BX648482.1 | - | 10 | 3443 |