Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320257.1 | NP_001307186.1 | 9 | 2363 | ||
NM_198460.2 | NP_940862.2 | 11 | 2704 | |||
XM_011540835.2 | XP_011539137.1 | 11 | 2533 | |||
GenBank mRNA | AK131329.1 | - | 6 | 1587 | ||
AK131356.1 | - | 11 | 2687 | |||
BX537949.1 | - | 11 | 2524 | |||
BX647907.1 | - | 11 | 2521 |