Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ITSN1 SIRNA Assays ›
Gene Symbol:
ITSN1
Gene Name:
intersectin 1
Gene Aliases:
ITSN, SH3D1A, SH3P17
Chromosome Location:
Chr.21: 33642400 - 33899861 on Build GRCh38
UniGene ID:
Hs.160324
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
12452
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ITSN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001132.1
NP_001001132.1
5
547
NM_003024.2
NP_003015.2
5
547
XM_005261026.2
XP_005261083.1
5
625
XM_005261027.1
XP_005261084.1
5
625
XM_005261031.1
XP_005261088.1
5
625
XM_011529691.1
XP_011527993.1
5
625
XM_011529692.1
XP_011527994.1
5
625
XM_011529693.2
XP_011527995.1
5
625
XM_017028428.1
XP_016883917.1
6
545
XM_017028429.1
XP_016883918.1
5
625
XM_017028430.1
XP_016883919.1
5
625
XM_017028431.1
XP_016883920.1
5
625
XM_017028432.1
XP_016883921.1
5
625
XM_017028433.1
XP_016883922.1
5
625
XM_017028434.1
XP_016883923.1
5
625
XM_017028435.1
XP_016883924.1
5
625
XM_017028436.1
XP_016883925.1
5
625
XM_017028437.1
XP_016883926.1
1
81
XM_017028438.1
XP_016883927.1
5
625
XM_017028439.1
XP_016883928.1
5
625
XM_017028440.1
XP_016883929.1
5
625
XM_017028441.1
XP_016883930.1
5
625
GenBank mRNA
AF064243.1
-
5
385
AF064244.1
-
5
385
AF114487.1
-
5
547
AF114488.1
-
5
547
AK027846.1
-
5
625
AK074554.1
-
5
483
AK075290.1
-
5
542
AK300274.1
-
5
522
BC013578.1
-
5
410
BC020269.1
-
5
460
BC039036.1
-
5
542
BC058925.1
-
5
574
BC073744.1
-
5
489
BC116185.1
-
5
358
40073780
BC116186.2
-
5
358
40073781
BC117560.1
-
5
527
7469144
BX648852.1
-
3
192
DQ340865.1
-
4
275
DQ386455.1
-
4
298
DQ679754.1
-
5
364
DQ679755.1
-
5
364
DQ679756.1
-
5
364
DQ679757.1
-
5
364
EU107386.1
-
5
364
EU117382.1
-
4
280
EU120733.1
-
4
280
EU120734.1
-
4
280
EU120735.1
-
5
364
EU120736.1
-
5
364
EU140799.1
-
5
364
EU140800.1
-
4
280
EU152331.1
-
4
280
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
small GTPase mediated signal transduction
regulation of Rho protein signal transduction
positive regulation of phosphorylation
positive regulation of apoptotic process
negative regulation of neuron apoptotic process
positive regulation of GTPase activity
ephrin receptor signaling pathway
synaptic vesicle endocytosis
regulation of small GTPase mediated signal transduction
positive regulation of protein kinase B signaling
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
calcium ion binding
protein binding
kinase activator activity
protein complex scaffold
proline-rich region binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-d4wl8:80/10.239.162.132:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0