Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: C3orf62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198562.2 | NP_940964.1 | 3 | 973 | ||
GenBank mRNA | AK096799.1 | - | 2 | 718 | ||
AK125642.1 | - | 3 | 917 | |||
BC023586.2 | - | 3 | 1807 | 4650297 | ||
BC053629.1 | - | 3 | 973 | 6204001 | ||
BC061697.1 | - | 3 | 963 | 6737728 |