Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: FAM47E-STBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242939.1 | NP_001229868.1 | 7 | 1424 | ||
GenBank mRNA | AK090914.1 | - | 5 | 897 | ||
AK092277.1 | - | 7 | 1403 | |||
AK316039.1 | - | 3 | 580 |
Gene Symbol: STBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003943.4 | NP_003934.1 | 2 | 1315 | ||
GenBank mRNA | AF062534.1 | - | 2 | 698 | ||
AK002155.1 | - | 1 | 219 | |||
AK123795.1 | - | 2 | 1315 | |||
BC022301.1 | - | 2 | 641 | 4694574 |