Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145713.1 | NP_001139185.1 | 7 | 1358 | ||
NM_001145714.1 | NP_001139186.1 | 8 | 1428 | |||
NM_005500.2 | NP_005491.1 | 9 | 1573 | |||
NR_027280.1 | 9 | 1622 | ||||
XM_006722963.2 | XP_006723026.1 | 10 | 1599 | |||
XM_017026136.1 | XP_016881625.1 | 9 | 1444 | |||
GenBank mRNA | AF046025.2 | - | 9 | 1509 | ||
AF090385.2 | - | 9 | 1518 | |||
AF161489.1 | - | 9 | 1521 | |||
AK021978.1 | - | 7 | 1352 | |||
AK027595.1 | - | 10 | 1695 | |||
AK055481.1 | - | 9 | 1538 | |||
AK090645.1 | - | 9 | 1591 | |||
BC000344.2 | - | 9 | 1526 | 2823028 | ||
BC003611.2 | - | 9 | 1494 | 2988252 | ||
BC018271.1 | - | 9 | 1533 | 3452340 | ||
CN386158.1 | - | 1 | 517 |