Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TRAIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005879.2 | NP_005870.2 | 6 | 560 | ||
XM_011533264.1 | XP_011531566.1 | 6 | 654 | |||
GenBank mRNA | AB451320.1 | - | 6 | 454 | ||
AB451449.1 | - | 6 | 454 | |||
AK292172.1 | - | 6 | 548 | |||
BC000310.2 | - | 6 | 541 | 2821007 | ||
BC019283.1 | - | 6 | 541 | 2821007 | ||
BX473682.1 | - | 6 | 568 | |||
U77845.1 | - | 6 | 556 |