Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL6IP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006407.3 | NP_006398.1 | 3 | 790 | ||
GenBank mRNA | AB097051.1 | - | 3 | 771 | ||
AF070523.1 | - | 3 | 796 | |||
AF115965.1 | - | 3 | 781 | |||
AF125530.1 | - | 3 | 782 | |||
AF161476.1 | - | 3 | 771 | |||
AK074612.1 | - | 3 | 742 | |||
AK223566.1 | - | 3 | 694 | |||
AK301637.1 | - | 4 | 704 | |||
AK303148.1 | - | 3 | 676 | |||
BC005143.2 | - | 3 | 713 | 3510436 | ||
BC020797.1 | - | 3 | 707 | 4134658 |