Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM107A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076778.2 | NP_001070246.1 | 4 | 1301 | ||
NM_001282713.1 | NP_001269642.1 | 4 | 1101 | |||
NM_001282714.1 | NP_001269643.1 | 5 | 1549 | |||
NM_007177.3 | NP_009108.1 | 5 | 1399 | |||
GenBank mRNA | AB023811.1 | - | 4 | 882 | ||
AF035283.1 | - | 4 | 1300 | |||
AF089853.1 | - | 4 | 1392 | |||
AF089854.1 | - | 5 | 1490 | |||
AK054720.1 | - | 5 | 1549 | |||
AK055443.1 | - | 3 | 1437 | |||
AK128791.1 | - | 4 | 928 | |||
AK298001.1 | - | 4 | 1056 | |||
AK316450.1 | - | 4 | 956 | |||
AL050264.1 | - | 4 | 914 | |||
BC010561.1 | - | 5 | 1000 | 4152118 | ||
EU569833.1 | - | 4 | 956 |