Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM107A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076778.2 | NP_001070246.1 | 4 | 1470 | ||
NM_001282713.1 | NP_001269642.1 | 4 | 1270 | |||
NM_001282714.1 | NP_001269643.1 | 5 | 1718 | |||
NM_007177.3 | NP_009108.1 | 5 | 1568 | |||
GenBank mRNA | AB023811.1 | - | 4 | 1051 | ||
AF035283.1 | - | 4 | 1469 | |||
AF089853.1 | - | 4 | 1561 | |||
AF089854.1 | - | 5 | 1659 | |||
AK054720.1 | - | 5 | 1718 | |||
AK055443.1 | - | 3 | 1606 | |||
AK128791.1 | - | 4 | 1097 | |||
AK298001.1 | - | 4 | 1225 | |||
AK316450.1 | - | 4 | 1125 | |||
AL050264.1 | - | 4 | 1083 | |||
BC010561.1 | - | 5 | 1169 | 4152118 | ||
EU569833.1 | - | 4 | 1125 |