Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF078859.1 | - | 11 | 1498 | ||
AF078868.1 | - | 11 | 1481 | |||
AF116703.2 | - | 8 | 1057 | |||
AF134478.1 | - | 11 | 1454 | |||
AK027523.1 | - | 11 | 1531 | |||
AK074710.1 | - | 10 | 1428 | |||
AK225112.1 | - | 11 | 1405 | |||
AK225138.1 | - | 11 | 1508 | |||
AL136546.1 | - | 10 | 1387 | |||
AL833441.1 | - | 11 | 1497 | |||
BC012842.1 | - | 11 | 1569 | 3861915 | ||
BC013925.1 | - | 8 | 1049 | 3685016 | ||
BC091522.1 | - | 11 | 1422 | 30513213 | ||
DQ250006.1 | - | 11 | 1423 |