Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244644.1 | NP_001231573.1 | 5 | 1500 | ||
NM_014043.3 | NP_054762.2 | 6 | 1592 | |||
XM_011533576.2 | XP_011531878.1 | 6 | 1423 | |||
GenBank mRNA | AK002165.1 | - | 6 | 1565 | ||
AK002180.1 | - | 6 | 1569 | |||
AL080122.1 | - | 6 | 1564 | |||
BC001553.1 | - | 6 | 1408 | 3460712 |