Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF638 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014972.2 | NP_001014972.1 | 2 | 1323 | ||
NM_001252612.1 | NP_001239541.1 | 2 | 1649 | |||
NM_001252613.1 | NP_001239542.1 | 2 | 1333 | |||
NM_014497.4 | NP_055312.2 | 2 | 1333 | |||
XM_011532768.2 | XP_011531070.2 | 2 | 1419 | |||
XM_017003809.1 | XP_016859298.1 | 2 | 1424 | |||
XM_017003810.1 | XP_016859299.1 | 2 | 1334 | |||
XM_017003811.1 | XP_016859300.1 | 2 | 1419 | |||
XM_017003812.1 | XP_016859301.1 | 2 | 1419 | |||
GenBank mRNA | AF534078.1 | - | 1 | 1013 | ||
AK098006.1 | - | 1 | 120 | |||
AK291198.1 | - | 2 | 1297 | |||
AK293938.1 | - | 2 | 1354 | |||
AK309221.1 | - | 2 | 1374 | |||
BC024000.1 | - | 2 | 1292 | |||
BC064530.1 | - | 1 | 1057 | |||
BC083513.1 | - | 2 | 1292 | 5540321 | ||
BC143728.1 | - | 2 | 1286 | 9052242 | ||
BX537425.1 | - | 2 | 1297 | |||
CR749322.1 | - | 2 | 1646 | |||
D83032.1 | - | 2 | 1325 |